Would you like to log in again?
Nalgene® 50 Platinum-Cured Silicone Tubing, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
8060-0060 8060-0020 8060-0260 8060-0100 8060-0200 8060-0120 8060-0080 8060-0050 8060-0130 8060-0040 8060-0070 8060-0140 8060-0190 8060-0310
16211-324CS 258.96 USD InStock
16211-324 63014-738 16211-334 16211-326 16211-342 63014-734 16211-320 16211-344 16211-332 16211-316 16211-338 63014-730 63014-732 16211-340
Nalgene® 50 Platinum-Cured Silicone Tubing, Thermo Scientific
Tubing Transfer Tubing
Flexible, durable, high-purity tubing is designed for a variety of pump and transfer applications, including those in the pharmaceutical, research laboratory, bioprocess manufacturing, food and beverage, and semiconductor industries.
  • Contains no phthalate plasticizers that can leach out into transported fluid
  • Durometer hardness: Shore A, 50
  • Autoclavable
Tubing resists adhesion of tissue, blood, proteins, and food products while imparting no taste or odor to the fluids. Manufactured with NALGENE® 50 silicone elastomer. Translucent, nontoxic, nonpyrogenic, and nonhemolytic.
Certifications: Complies with USDA, USP Class VI, and 3A standards.
Ordering information: Supplied in 50' coil lengths.
Caution: Note: Not intended for invasive use.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty