Would you like to log in again?
NALGENE® Polypropylene Screw Closures, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
712171-0530 712174-0530 712150-0240 712150-0280 712150-1000 712150-0530 712150-0630 712150-0430
16219-150PK 78.41 USD InStock
16219-150 16219-030 16219-041 16219-080 16219-106 16219-301 16219-104 16219-102
NALGENE® Polypropylene Screw Closures, Thermo Scientific
Closures Bottle Closures
Linerless closures are for use only with NALGENE® narrow- and wide-mouth bottles and Erlenmeyer flasks
Note: Before autoclaving, set cap on top of container without engaging threads.
Autoclavable. Available in natural and amber.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty