Would you like to log in again?
NALGENE® Polypropylene Screw Closures, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
712160-0530 712160-0384 712160-0830
16225-234PK 118.48 USD InStock
16225-234 16225-204 16219-122
NALGENE® Polypropylene Screw Closures, Thermo Scientific
Closures Bottle Closures
Caps for large NALGENE® bottles and carboys
Note: Before autoclaving, set cap on top of container without engaging threads.
Caps 16225-204 and -234 are buttress-style. Autoclavable.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty