Would you like to log in again?
Nalgene® Closures with Barbed Bulkhead Fittings, White PP, Thermo Scientific
Supplier: Thermo Scientific
DS2167-0002 DS2167-0001
16225-393CS 160.23 USD InStock
16225-393 16225-391
Nalgene® Closures with Barbed Bulkhead Fittings, White PP, Thermo Scientific
Closures Bottle Closures
Thermo Scientific Nalgene Closures with Barbed Bulkhead Fittings support the integration of tubing sets to Nalgene culture vessels.
Fittings may also be used with any Nalgene container with a 38-430 sized closure. The 38-430 closures are pre-drilled to accept either a 1/4 or 1/2 inch (6-13mm) ID fitting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty