Would you like to log in again?
B.O.D. Bottles with Black PVC Coating, 300 ml, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
227667
16285-602CS 716.58 USD InStock
16285-602
B.O.D. Bottles with Black PVC Coating, 300 ml, WHEATON®, DWK Life Sciences
Bottles BOD Bottles
Ideal for light sensitive samples.
  • Coated with black PVC plastic to block all visible light up to 800 nm to inhibit the production of oxygen by algae
  • Recommended for use in marine photosynthesis projects when comparing oxygen in a light and a dark bottle
  • Manufactured from USP Type I borosilicate glass
Delivery information: Supplied with a glass robotic stopper and an opaqueblack cap.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty