Would you like to log in again?
Serum Bottle, Polypropylene, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
224007
16300-053CS 381.35 USD InStock
16300-053
Serum Bottle, Polypropylene, WHEATON®, DWK Life Sciences
Bottles Plastic Bottles
Serum bottles are designed for use with diluent solutions.
Not recommended for IV or other solutions which require a glass container.
They are autoclavable at 121 °C (250 °F) for 20 min.
Ordering information: This bottle accepts rubber stoppers and aluminum seals.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty