Would you like to log in again?
Serum Bottle, High Density Polyethylene, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
224031 224033 224037 224036
16300-056CS 362.79 USD InStock
16300-056 16300-057 16300-058 16300-059
Serum Bottle, High Density Polyethylene, WHEATON®, DWK Life Sciences
Bottles Plastic Bottles
These serum bottles are manufactured from high density polyethylene and are designed for use in the storage of non-medical products.
Ordering information: These bottles accept standard rubber stoppers and aluminum seals.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty