Would you like to log in again?
WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
242411-01 242402 242423 W242417-A W242407-A W242426-A 242401 242404 242438 242412 W242425-A W242436-A W242420-A W242406-A 242421-01 W242404-A W242415-A 242431-01 W242433-A W242430-A W242411-A 242401-01 242427 242410 242433 W242431-01-A W242401-01-A W242428-A W242427-A W242422-A W242440-A W242421-A 242414 242417 W242421-01-A W242411-01-A W242403-A W242432-A W242418-A W242412-A 242406 W242437-A W242401-A 242426 W242402-A 242422 242411 W242423-A 242425 242405 W242405-A W242438-A 242407 W242414-A W242408-A 242408 242413 242415 242421 W242416-A W242434-A 242430 W242424-A 242416 W242410-A 242440 242420 242436 242434 242437 W242413-A 242432 242403 242431 W242431-A 242424
89220-238CS 397.77 USD InStock
89220-238 16300-087 16300-088 16300-089 89220-234 89220-276 89220-230 89500-066 89500-064 16300-081 16300-082 89500-062 16300-084 16300-085 89500-060 16300-086 80086-304 89500-068 89220-208 89220-206 89220-202 89220-246 16300-098 16300-099 89220-288 89220-242 89220-284 89500-054 89220-280 16300-092 16300-093 16300-094 16300-095 16300-096 89500-058 89500-056 89220-216 89500-080 16300-065 16300-066 16300-100 16300-067 89220-212 16300-068 89220-256 89048-956 16300-069 16300-102 89220-210 89220-254 89220-250 89500-086 89500-084 16300-062 16300-063 89500-082 16300-064 89220-226 89220-224 16300-076 89220-268 16300-077 16300-078 89220-220 89220-264 89220-260 89500-076 89048-954 16300-071 89500-074 16300-072 16300-073 89500-072 16300-074 89500-070 89500-078
WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
Closures Bottle Closures
Extended Controlled Dropper Tips are made of LDPE (Low-Density Polyethylene)
Extended Controlled Dropper Tips snap into the inside of the bottle neck and are available in bulk. Dropper tips can be covered with Wheaton dropper tip screw caps.
They dispense one drop at a time. The average drop size is 44µL (±6µL).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty