Would you like to log in again?
Hybridization Bottles, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
805027 805031 805020 805000 805021 805023 805003 805011 805007 805001
16300-304EA 96.68 USD InStock
16300-304 16300-316 16300-306 16300-318 16300-308 16300-320 16300-300 16300-312 16300-302 16300-314
Hybridization Bottles, WHEATON®, DWK Life Sciences
Bottles Hybridization Bottles/Tubes
These bottles are ideal for nucleic acid and protein blotting procedures that use a minimum of probe solution.
These borosilicate bottles are produced with a heavy wall for added safety and are available with a slip-resistant safety coating. The 35 mm bottles incorporate a fluoropolymer resin/silicone lined cap to prevent leakage. Both bottles incorporate a polypropylene cap with a welded fluoropolymer resin/silicone liner. Optional acrylic storage rack holds up to six hybridization bottles.
They are a safe and efficient alternative to messy bags, and allow hybridization and wash solutions to be quickly poured in and out. Bottles fit most popular hybridization ovens to simplify procedures such as Southern, Northern, Western, and slot blots.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty