Would you like to log in again?
Nalgene® Carboys with Spigot and Handles, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2318-0020 2318-0065 2318-0010 2318-0130 2318-0050
16334-184CS 878.8 USD InStock
16334-184 16334-209 16334-242 16334-264 16334-220
Nalgene® Carboys with Spigot and Handles, Low-Density Polyethylene, Thermo Scientific
Bottles Carboys
Suitable for collecting and dispensing distilled water, reagents, and acids.
  • Leakproof
Spigots accept 16 mm (⁵/₈") Int.Ø tubing, and include an HDPE tubing adapter that takes tubing with 6 to 8 mm (¼ to ⁵/₁₆") Int.Ø. The ETFE spigot is for use with applications with selected solvents and chemicals in which polypropylene spigots are unsatisfactory.
Graduated to deliver in 1 gal. and 5 L increments. Spigot boss molded in one piece with bottle, not welded on, for strength and leak resistance. Spigot comes with adapter to accommodate a variety of tubing sizes. The 3.8 L (1 gal.) size does not have handles.
Ordering information: Supplied with carboy, polypropylene spigot, and screw cap. For replacement caps, see 16219-122 series.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty