Would you like to log in again?
Dropping Bottle, with Tip and Cap, LDPE, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
211632 211634 211621 211635 211623 211630 211631 211625 211620 211622 211624 211633
16347-072CS 521.89 USD InStock
16347-072 16347-050 16347-070 16347-028 16347-026 16347-024 16347-022 16347-054 16347-032 16364-980 16347-074 16347-052
Dropping Bottle, with Tip and Cap, LDPE, WHEATON®, DWK Life Sciences
Bottles Dropping and Dispensing Bottles
WHEATON Dropping Bottles are made of LDPE (Low Density Polyethyleye)
The extended controlled dropper tip dispneses one drop at a time, with an average drop size of 44 µl ± 6 µl. Radiation grade polypropylene screw caps and tips packaged separately. Dropping bottles, natural tips and white caps are packaged separately.
Dropping bottles with tips can be used to dispense a stream of liquid or a non-controlled drop. Dropping bottles, tips and caps are packaged separately and bottles are autoclavable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty