Would you like to log in again?
Nalgene® Drop-Dispenser Bottle, Teflon® FEP, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2414-0030
16354-320CS 442.73 USD InStock
16354-320
Nalgene® Drop-Dispenser Bottle, Teflon® FEP, Thermo Scientific
Bottles Dropping and Dispensing Bottles
Made of Teflon® FEP and ETFE dropping closure and cap. Transparent bottle can be used with virtually any chemical. Contamination-free dispensing. Repeatedly autoclavable with tip uncapped.
Convenient one-hand operationjust squeeze bottle to dispense. Dispenses one drop at a time.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty