Would you like to log in again?
Nalgene® Dispensing Bottles, Polypropylene Copolymer, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
DS2420-0250 DS2420-0125
16354-739CS 109.44 USD InStock
16354-739 16354-706
Nalgene® Dispensing Bottles, Polypropylene Copolymer, Thermo Scientific
Bottles Dropping and Dispensing Bottles
Produce a fine steady stream but only when squeezed. Linerless, noncontaminating 24-415 closure with captive spout cap. Leakproof. Bottles may be autoclaved full, with tips uncapped.
For dispensing sterile solutions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty