Would you like to log in again?
Laboratory Bottles, Wide Mouth, HDPE, Dynalon
Supplier: Dynalon
301605-0001 301605-0016 301605-0008 301605-0004 301605-0032
16354-824PK 40.78 USD InStock
16354-824 30620-180 16354-828 47743-012 30620-178
Laboratory Bottles, Wide Mouth, HDPE, Dynalon
Bottles Plastic Bottles
High density polyethylene wide mouth lab bottles are leakproof, rigid, and translucent, and offer excellent chemical resistance.
Supplied with a polypropylene closure, they are ideal for storage and handling of liquids or solids. Manufactured from FDA approved resin for non-contaminating storage of chemicals and food grade products.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty