Would you like to log in again?
Laboratory Bottles, Wide Mouth, Polypropylene, Dynalon
Supplier: Dynalon
301625-0008 301625-0032 301625-0002
30620-194CS 215.4 USD InStock
30620-194 16354-836 30620-190
Laboratory Bottles, Wide Mouth, Polypropylene, Dynalon
Bottles Plastic Bottles
Leakproof, rigid, and translucent wide mouth polypropylene lab bottles can be autoclaved.
  • Excellent chemical resistance
  • Ideal for storage and sampling of liquids or solids
Delivery information: Supplied with a polypropylene closure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty