Would you like to log in again?
Reservoir Bottle with Bottom Valve Outlet, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
14612F-10000 14612F-5000 14612F-2000 14612F-20000
16429-526CS 1744.32 USD InStock
16429-526 16429-524 16429-522 16429-520
Reservoir Bottle with Bottom Valve Outlet, Kimble Chase, DWK Life Sciences
Bottles Solution Bottles
Designed to store and discharge liquids via a bottom valve outlet.
KIMAX® bottles are graduated with an approximate capacity scale. Bottom outlet is controlled with an 8 mm valve which can be used with ¹/₄" flexible tubing attached to the quick-release hose barb connector and with rigid ¹/₄" Ext.Ø PTFE tubing with the o-ring and cap supplied on the quick-release connector.
Certifications: Ref: ASTM Method D1744. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty