Would you like to log in again?
Solution Bottle with Narrow Mouth for Rubber Stopper, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
15093-20000 15093-1000 15093-10000 15093-5000 15093-2000
16429-638CS 479.11 USD InStock
16429-638 16429-636 16429-644 16429-642 16429-640
Solution Bottle with Narrow Mouth for Rubber Stopper, Kimble Chase, DWK Life Sciences
Bottles Solution Bottles
Ideal for storage and dispensing of solutions.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Neck is tooled for increased mechanical strength and for a uniform fit with rubber stoppers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty