Would you like to log in again?
F-Style Cans, Metal
Supplier: Qorpak
MET-03045 280593 MET-03040 MET-05426 MET-03043
76539-372CS 658.03 USD InStock
76539-372 16455-234 76539-368 76406-944 16455-232
F-Style Cans, Metal
Safety Cans
Metal F-Style Oblong cans are ideal for products such as cooking oils, paint thinners, turpentine, lighter fluids, insecticides, and general storage applications.
They are compact and easily fit on a shelf. Screw top lids available on oblong cans provide for packaging liquids or pourable emulsions. Air tight seal is provided at the time of original filling and can readily be achieved when resealing after use. This sealing arrangement is used when it is desirable to be able to pour product, or to apply small quantities to a cloth or sponge. May be sterilized by heat if necessary. Capable of withstanding a great deal of physical shock in shipping and storage. Compatible with a wide range of products and temperatures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty