Would you like to log in again?
TAPE 3M 371 CLEAR 48MMX100M
Supplier: XPEDX-DALLAS
3M* Scotch* Sealing Tape^ Clear. Width: 5.1cm. Length: 100m. Pressure-sensitive rubber resin adhesive. Biaxially oriented polypropylene backing is easy to handle and dispense. Resists abrasion, moisture, chemicals, and scuffing. Conforms around edges and over rough surfaces for a tight seal.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TAPE 3M 371 CLEAR 48MMX100M 16510-526 10036270 Case of 36 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty