Would you like to log in again?
VWR® Wide Mouth Wash Bottles, LDPE
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
00486-0500 00484-0125 00484-0250 00485-0250 00486-0250 00484-1000 00487-1000 00487-0500 00485-1000 00487-0125 00487-0250 004860125 00486-1000
16650-333CS 141.57 USD InStock
16650-333 16650-322 16650-355 16650-344 16650-107 16650-093 16650-082 16650-264 16650-242 16650-173 16650-195 16650-184 16650-162
VWR® Wide Mouth Wash Bottles, LDPE
Bottles Wash Bottles
These bottles save time, help avoid mistakes, and increase safety.
  • Color-coded caps provide quick, safe identification
  • Wide-mouthed design
  • One-piece cap and nozzle plus delivery tube
  • Single drop or needle-thin stream
Color-coded, wide mouth bottles are ideal for the quick identification of the many special solutions and reagents needed in your lab. Bottles are durable, chemical-resistant, squeezable, and easy to clean. One-piece colored cap and nozzle are polypropylene. Delivery tube is polyethylene. The leakproof wash cap assembly will deliver a single drop or a needle thin stream, with finger-tip squeeze control of flow. For a coarser flow, just cut the nozzle tip back.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty