Would you like to log in again?
Nalgene® Economy Wash Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2401-0250 2401-0125 2401-0500 2401-1000
16651-518CS 108.48 USD InStock
16651-518 16651-450 16651-471 16651-493
Nalgene® Economy Wash Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Wash Bottles
Through-the-top wash bottles feature closure and stem molded in one piece for leakproof service.
Draw tube is welded into closure to prevent separation. The tip dispenses a uniform stream, and can be cut back to increase flow. Supplied with polypropylene screw cap and stem.
Caution: Not recommended for long-term use with acetone.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty