Would you like to log in again?
Wash Bottle, Easy Squeeze, Low Density Polyethylene, Dynalon
Supplier: Dynalon
224115 224085
16651-631CS 328.19 USD InStock
16651-631 16651-620
Wash Bottle, Easy Squeeze, Low Density Polyethylene, Dynalon
Bottles Wash Bottles
Easy-squeeze wash bottles with integrally molded tube.
Requires only gentle pressure to eject and accurately direct the liquid. The dispensing tip can be cut back to increase the flow rate. Maximum temperature of LDPE is 80 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty