Would you like to log in again?
Nalgene® Wash Bottles, Low-Density Polyethylene, Wide Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2407-1000 2407-0500
16651-664CS 134.4 USD InStock
16651-664 16651-642
Nalgene® Wash Bottles, Low-Density Polyethylene, Wide Mouth, Thermo Scientific
Bottles Wash Bottles
Wide mouth design for easy filling.
Through-the-top polypropylene screw closure and stem molded in one piece. Tip can be cut back to increase flow. Leakproof.
Caution: Not recommended for long-term use with acetone.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty