Would you like to log in again?
Nalgene® Unitary™ Safety Wash Bottles, Thermo Scientific
Supplier: Thermo Scientific
DS2408-0250 DS2408-0500
16651-961CS 93.02 USD InStock
16651-961 16651-949
Nalgene® Unitary™ Safety Wash Bottles, Thermo Scientific
Bottles Wash Bottles
Nalgene® Unitary™ Safety Wash Bottles are molded in one piece which eliminates possible contamination from a delivery tube and offers safety from chemical splashes when removing the closure.
The bottles also feature a leakproof linerless polypropylene closure, and are red for easy identification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty