Would you like to log in again?
KIMAX® Cylindrical Weighing Bottles, Regular and Tall Form, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
15145-3060 15145-4050 15145-1550 15145-2540 15145-2550 15145-4080 15145-5060
16711-121CS 637.71 USD InStock
16711-121 16711-143 16711-100 16711-167 16711-063 16711-085 16711-020
KIMAX® Cylindrical Weighing Bottles, Regular and Tall Form, Kimble®, DWK Life Sciences
Bottles Weighing Bottles
Bottles are supplied with a plug-style, closed bottom airtight glass [ST] stopper that fits into an internally ground body.
Bottles also feature a frosted marking area. Designed from Federal Specification NNN-B-1493, Type I, Style I requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty