Would you like to log in again?
KIMAX® Cylindrical Weighing Bottles, Low Form, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
15165-5030 15165-7033 15165-6030
16716-023CS 405.32 USD InStock
16716-023 16716-067 16716-045
KIMAX® Cylindrical Weighing Bottles, Low Form, Kimble®, DWK Life Sciences
Bottles Weighing Bottles
Supplied complete with closed bottom stopper ground into the neck.
Designed from Federal Specifications NNN-B-1493, Type I, Style I. Capacity indicates measurement to base of neck.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty