Would you like to log in again?
HyClone™ Fetal Bovine Sera Defined (United States), HyClone Products (Cytiva)
Supplier: HyClone products (Cytiva)
HyClone™
SH30070.02HI SH30070.01 SH30070.03 SH30070.02 SH30070.03HI SH30070.02 SH30070.02HI SH30070.01 SH30070.03HI SH30070.03
16777-530EA 1398.48 USD InStock
16777-530 16777-002 CA82013-568 16750-030 CA16750-030 CA16777-006 16777-006 82013-568 CA16777-002 CA16777-530
HyClone™ Fetal Bovine Sera Defined (United States), HyClone Products (Cytiva)
Sera (Serum)
Defined foetal bovine serum (FBS) is the highest quality of FBS available and is very widely used by those cell culturist who have a concern for viral contaminants and require an extensive biochemical profile. Defined FBS is filtered through multiple 40 nm (0.04 µm) pore-size related filters, which are the most retentive filters used in commercial FBS production. Over 50 components are analysed and the results are included on the biochemical assay.
  • Filtered through serial 40 nm pore-size rated filters
  • Maximize cell culture growth
  • U.S. sourced
Maximize cell culture growth with U.S.-sourced Cytiva HyClone Defined Fetal Bovine Serum (FBS), filtered through serial 40nm pore-size rated filters and tested for endotoxin, hemoglobin, and extensive biochemical profile assays.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty