Synonyms:
Dihydrogen oxide
SH30538.FS
SH30538.01
SH30538.FS
SH30538.02
SH30538.LS
SH30538.03
SH30538.01
SH30538.03
SH30538.LS
SH30538.02
82007-332EA
18.47
USD
InStock
82007-332
CA16750-128
82007-336
CA82007-334
82007-334
CA82007-332
CA82007-336
16750-104
CA16750-104
16750-128
Water, HyClone™ HyPure, Molecular Biology Grade
Water
Nuclease-free HyClone Molecular Biology-Grade Water, containing no DEPC or other carcinogenic chemicals is used for critical applications involving nucleotides such as RNA and DNA.
- Significantly low levels of endotoxins
- Certified to have no detectable activity in DNase, RNase, and protease assays
- Maximum allowable limit of endotoxin is 0.05 EU/mL
HyClone HyPure Molecular Biology Grade Water purification process includes continuous deionization, reverse osmosis, ultraviolet light treatment, distillation, and 0.1 µm filtration.
Certifications: Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP).filtration