Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, HyClone™ HyPure, Cell Culture Grade

Supplier: Cytiva

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Dihydrogen oxide

SH30529.01 SH30529.LS SH30529.03 SH30529.01 SH30529.LS SH30529.02 SH30529.FS SH30529.02 SH30529.03 SH30529.FS
82007-330CS 186.72 USD InStock
CA82007-330 82007-330 82007-326 16750-098 CA82007-328 CA82007-326 82007-328 CA16750-098 CA16750-114 16750-114
Water, HyClone™ HyPure, Cell Culture Grade
Water
Eliminate endotoxin concerns with endotoxin-free HyClone Cell Culture-Grade Water, available in a variety of volumes for both research and manufacturing processes.

  • Undergoes extensive purification to meet or exceed the stringent specifications of United States Pharmacopeia (USP)
  • Meets < 0.005 EU/mL endotoxin by turbidimetric assay
  • Manufactured in our ISO 9001 certified facilities

HyClone HyPure Cell Culture Grade Water can be used in multiple cell culture applications. With endotoxin levels as low as < 0.005 EU/mL, HyPure Cell Culture Grade Water supports healthy cell growth and consistent experimental results.

Certifications: Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP).


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Order Now
Specification Test Results
Endotoxin Concentration < 0.005 EU/mL
Sterilization 0.1 µm sterile filtered
pH 5.0 to 7.0
Storage 2° to 30° C

 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty