Would you like to log in again?
HyClone™ DMEM/F12 1:1 Cell Culture Media, Cytiva
Supplier: HyClone products (Cytiva)
HyClone™
SH30004.04 SH30004.04
16777-056EA 363.42 USD InStock
16777-056 CA16777-056
HyClone™ DMEM/F12 1:1 Cell Culture Media, Cytiva
Cell Culture Media
HyClone™ DMEM/F12 is an equal parts blend of DMEM and Ham’s F12 media that is widely used for culturing many different mammalian cells including MDCK, Glial cells, fibroblasts, and endothelial cells. This formulation combines the high concentration of glucose, amino acids and vitamins from DMEM with the wide variety of other components available in Ham’s F-12.
  • Contains 17,5 mM glucose and 0,5 mM sodium pyruvate
  • Available with or without L-glutamine or HEPES
  • Available in multiple sizes and configurations of liquid or powder
  • All liquid media products are hydrated using WFI quality water
  • 0.1 μm sterile filtered
Manufactured using ISO 9001:2000-certified processes.
ADCF=Animal derived component free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty