Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
HyClone™ Ham's Nutrient Mixture F10/F12, Cell Culture Media, Powdered, Cytiva
Supplier: HyClone products (Cytiva)
HyClone™
SH30010.03 SH30010.04 SH30010.02 SH30010.04 SH30010.02 SH30010.03
16777-089EA 71.63 USD InStock
16777-089 CA16777-092 16777-092 CA16777-090 16777-090 CA16777-089
HyClone™ Ham's Nutrient Mixture F10/F12, Cell Culture Media, Powdered, Cytiva
Cell Culture Media
Dry powdered media for supporting cell culture growth and biomanufacturing processes.
  • Raw material components screened through strict quality control testing for lot-to-lot consistency.
  • Products are hydrated using purified process water and undergo sterile filtration.
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.
Caution: Storage 2° to 8°C, away from light
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty