Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HyClone™ Dulbecco's Phosphate Buffered Saline, Powder, HyClone products (Cytiva)

Supplier: HyClone products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

HyClone™
SH30013.04 SH30013.02 SH30013.02 SH30013.03 SH30013.03 SH30013.04
16777-105EA 81.67 USD InStock
CA16777-105 CA16777-102 CA16777-103 16777-105 16777-102 16777-103
HyClone™ Dulbecco's Phosphate Buffered Saline, Powder, HyClone products (Cytiva)
Buffers Cell Culture Buffers
Balanced salt solution used for multiple cell culture applications (powder form).

  • Formulated and processed for rapid dissolution
  • Non sterile
  • Pre-weighed powders ready for hydration to 1X
  • Available in 3" ported funnel shaped containers upon request
  • Available in lot sizes up to 6500 kg.

For making isotonic solutions for cell or tissue rinsing, cell dilution, reagent prep, transportation etc. where maintanence of cell integrity is required for further manufacturing or research use.

Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.

Caution: Not for diagnostic or therapeutic use.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty