Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydrogen carbonate 75 g/l in aqueous solution, HyClone™, sterile filtered

Supplier: Cytiva
Synonyms: Sodium bicarbonate

SH30033.01 SH30033.01
16777-160EA 17.69 USD InStock
CA16777-160 16777-160
Sodium hydrogen carbonate 75 g/l in aqueous solution, HyClone™, sterile filtered
Sodium hydrogen carbonate
Buffer cell culture with a fully QC-tested solution.

  • 0.1 µm sterile filtered
  • pH 7.9 to 9.4
  • Endotoxin Concentration: ≤1.0 EU/mL

This solution has full traceability and documented origin of all formula ingredients. It is validated and consistent for production processes.

Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.


Formula: NaHCO₃
MW: 84.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/81/16640581.jpg
Order Now
Specification Test Results
Endotoxin concentration ≤1.0 EU/mL
Sterilization 0.1 µm sterile filtered
pH 7.9 - 9.4

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty