Would you like to log in again?
HyClone™ Amino Acids, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
SH30034.02 SH30034.01 SH30034.02 SH30034.01
82007-322EA 45.93 USD InStock
82007-322 CA16777-162 16777-162 CA82007-322
HyClone™ Amino Acids, HyClone products (Cytiva)
Cell Culture Supplements and Additives
HyClone Amino Acids are ideal for supplementation of serum- and protein-free media
They support the growth of cells that have high energy demands and synthesize large amounts of proteins and nucleic acids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty