Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HyClone™ Dulbecco's Phosphate Buffered Saline Solution, HyClone products (Cytiva)

Supplier: HyClone products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SH30028.LS SH30028.FS SH30028.01 SH30028.03 SH30378.02 SH30378.02 SH30028.08 SH30028.04 SH30264.FS SH30028.02 SH30264.02 SH30028.02 SH30028.04 SH30378.03 SH30264.01 SH30028.03 SH30028.01 SH30028.LS SH30028.08 SH30264.01 SH30378.03 SH30028.FS SH30264.FS SH30264.02
82013-222EA 9.94 USD InStock
82013-222 CA82013-318 CA16750-110 16750-110 16750-102 CA16750-076 CA95059-620 CA16750-102 95059-620 CA82020-066 CA16777-150 16750-076 16777-150 CA82024-294 82024-294 82013-318 CA16777-644 CA16777-259 16777-644 82020-066 CA16777-257 16777-259 CA82013-222 16777-257
HyClone™ Dulbecco's Phosphate Buffered Saline Solution, HyClone products (Cytiva)
Buffers Cell Culture Buffers
Balanced salt solution used for multiple cell culture applications, (liquid form).

  • Certificate of Analysis included with product and available electronically for future reference.
  • Standard stock items from 100 mL to 200 L
  • Volumes up to 1000 L available as made to order
  • Available in 1X and 10X concentrations to meet specific needs

For making isotonic solutions for cell or tissue rinsing, cell dilution, reagent prep, transportation etc. where maintanence of cell integrity is required for further manufacturing or research use.

Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.

Caution: Not for diagnostic or therapeutic use.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty