Would you like to log in again?
HyClone™ USDA Import Tested Fetal Bovine Serum (FBS), Cytiva
Supplier: HyClone products (Cytiva)
HyClone™
SH30910.03HI 3091003IR25-40 SH30910.02HI SH30910.03 SH30910.03 SH30910.03 SH30910.02HI SH30910.03HI SH30910.02 SH30910.02 3091003IR25-40
76324-916EA 471.79 USD InStock
76324-916 89133-100 89133-098 89133-096 CA89133-094 16777-538 89133-094 CA89133-096 CA89133-098 CA89133-100 CA76324-916
HyClone™ USDA Import Tested Fetal Bovine Serum (FBS), Cytiva
Sera (Serum)
HyClone USDA-tested Fetal Bovine Serum (FBS) provides important proteins, hormones, growth factors, metabolites, and nutrients for your cell culture processes at a competitive price. USDA-tested FBS is ideal for typical cell culture research applications.
  • Complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Produced using true pool technology for uniformity and consistency from bottle to bottle
  • Filtered through three sequential 100 nm (0,1 µm) pore size-rated filters
  • Sourced from countries recognized as being low risk of BSE and FMD by the USDA
  • Once thawed, sera should be stored at 2 to 8 °C
USDA-Tested Fetal bovine serum (FBS) is sourced from bovine spongiform encephalopathy (BSE)-negligible regions in throughout the world for use in low-risk applications. This serum is filtered in the US and considered to be foot and mouth disease (FMD)-free.
Each lot is safety tested by the U.S. Department of Agriculture (USDA) in accordance with safety protocols required for the serum country of origin. This serum is largely sourced from Central America.
Additional treatments available including heat-inactivation and irradiation. Additional testing, including EMEA-conforming testing, is also available.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty