Would you like to log in again?
Chart Papers, Standard, Omega Engineering
Supplier: Omega Scientific
CT485-CMF CT485-CDF CT485-CMC CT485-CWF
17000-008PK 151.07 USD InStock
17000-008 17000-016 17000-014 17000-010
Chart Papers, Standard, Omega Engineering
Chart Recorders Charts for Recorders
These standard double sided chart papers are compatible with CTXL series of tempearture and relative humidity recorders.
Packaging: Single package of 100 charts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty