Would you like to log in again?
Cylinder and Bottle Brush, Mill-Rose
Supplier: Mill-Rose
72102B
17041-002PK 47.76 USD InStock
17041-002
Cylinder and Bottle Brush, Mill-Rose
Brushes
Thick natural black bristles are densely packed for thorough cleaning of cylinders, jars and bottles.
The tufted end facilitates cleaning the bottom. Natural black bristle in a twisted wire handle.
Tip (tufted end) diameter: 4.5 cm (1.75")
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty