Would you like to log in again?
Babcock Milk Test Bottle Brush, Mill-Rose
Supplier: Mill-Rose
72222B
17068-009PK 65.69 USD InStock
17068-009
Babcock Milk Test Bottle Brush, Mill-Rose
Brushes
Specially sized for the extra narrow neck of Babcock milk test bottles.
  • Short dense bristles along the stem provide close scrubbing to help remove butterfat residue
  • Natural black bristles twisted into a wire handle with hanging loop
Extra long natural bristles fold to fit through the narrow bottle neck then expand for washing the bottle body.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty