Would you like to log in again?
Test Tube Brushes, Tie Tip, Justman
Supplier: Justman Brush
J610112
17140-000EA 24.6 USD InStock
17140-000
Test Tube Brushes, Tie Tip, Justman
Brushes
Tied tip brushes are designed to enter narrow mouthed bottles and tubes and then expand to clean the sides of the bottle and tube, while protecting the bottom of the bottle or tube from being scratched by the wire tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty