Would you like to log in again?
Test Tube Brushes, Justman
Supplier: Justman Brush
J610132 J610164 J710122 J610156 J610256 J710158 J710150
17145-080EA 13.41 USD InStock
17145-080 17140-006 17145-005 30620-538 17145-002 30620-550 17080-020
Test Tube Brushes, Justman
Brushes
Test tube brush is especially designed to clean the inner walls and bottom of test tubes to remove every trace of the chemical stored in it.
  • White-colored bristle with silver handle
  • Test tube brush with a radial tip
The tip of this brush features formed sprays that thoroughly clean the bottom of the test tube without damaging or scratching the glassware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty