Would you like to log in again?
SP Bel-Art Metal Quick Disconnect Couplings, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F19724-0204 F19723-0802 F19724-0804 F19723-0202
17150-050EA 66.95 USD InStock
17150-050 17150-052 17150-054 17150-056
SP Bel-Art Metal Quick Disconnect Couplings, Bel-Art Products, a part of SP
Fittings and Connectors
Chrome plated brass connectors with Buna-N O-Ring (made by Colder Products) feature easy-to-use slide locks for leak-free, in-line connections, and automatic shut off when disconnected.
  • Easy-to-use design
  • Leakproof in-line connections
  • Automatic shutoff feature when disconnected
  • Barbed ends on hose
Hose barb ends accept flexible tubing.
Operate at temperatures to 82°C (180°F), pressures to 1.7Mpa (250psi), and vacuum to 30 microns of Hg. Chrome plated valves provide dependable operation and are ideal for liquid and gas connectors in biomedical instrumentation and general industrial applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty