Would you like to log in again?
Cylinder and Funnel Brush, Mill-Rose
Supplier: Mill-Rose
72121B
17160-001PK 104.87 USD InStock
17160-001
Cylinder and Funnel Brush, Mill-Rose
Brushes
Specially designed for cleaning conical glassware, such as pharmaceutical graduates and funnels.
  • Bristles are held with twisted wire
  • Smooth wooden handle
Tapered black bristles have a rounded bristle tip and double tuft extensions for easy removal of residues.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty