Would you like to log in again?
Imhoff Cone Brush, Mill-Rose
Supplier: Mill-Rose
72301B
17161-004EA 62.6 USD InStock
17161-004
Imhoff Cone Brush, Mill-Rose
Brushes
Tapered brush head flares out in a fan tip for increased scrubbing area when removing stubborn sediments.
  • Natural black bristles and vegetable fiber are wired to form a dense effective cleaning tool
  • Round smooth wooden handle
Brush taper: 1.9–10.2 cm (3/4–4")
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty