Would you like to log in again?
Beaker and Jar Brushes, Mill-Rose
Supplier: Mill-Rose
MR-75010W MR-75001B MR-75002W
17181-001EA 49.33 USD InStock
17181-001 17180-009 17176-023
Beaker and Jar Brushes, Mill-Rose
Brushes
Designed for heavy-duty cleaning of beakers, jars, and other wide-mouth glassware.
  • Heavy tufting at the tip reduces scratching and improves cleaning capability
  • Handle has an opening for hanging
Long-lasting bristles are spirally wound into grooves in the smooth wood handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty