Would you like to log in again?
Beaker, Jar, and Cylinder Brushes, Mill-Rose
Supplier: Mill-Rose
72112B 72110B 2111B
17170-005EA 40.52 USD InStock
17170-005 17190-007 17190-002
Beaker, Jar, and Cylinder Brushes, Mill-Rose
Brushes
Completely rounded tip provides safe, effective cleaning for wide mouth glassware such as beakers, jars, and cylinders.
  • Extra dense natural black bristles are mounted on twisted wires into a durable wood handle
  • Double-tuft extensions at the tip for reaching into corners
  • Available with two or four rows of bristles
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty