Would you like to log in again?
Camel Hair Brushes, Mill-Rose
Supplier: Mill-Rose
MR75901-6 MR75901-3
17216-048PK 51.2 USD InStock
17216-048 17216-026
Camel Hair Brushes, Mill-Rose
Brushes
Soft tapered brushes, originally designed for the fine arts, are excellent for dusting balances and delicate laboratory equipment.
Natural, washable camel hair is attached to hardwood handles by metal ferrules.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty