Would you like to log in again?
SP Bel-Art Small Pails, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
SP Bel-Art
F16775-0000 F16771-0000
17308-050EA 72.87 USD InStock
17308-050 70690-030
SP Bel-Art Small Pails, Bel-Art Products, a part of SP
Buckets
These 7.6 liter (8qt.) plastic pails are available in polyethylene or autoclavable polypropylene.
  • Polyethylene or polypropylene construction
  • Polypropylene model is autoclavable
  • Snug-fitting lid
  • Removable plated steel support ring
They have a snug-fitting lid and removable plated steel support ring and bail handle. Bucket height is 25 cm (10"), 20 cm (8") I.D. at top. Polypropylene pail and cover 17308-050 are steam autoclavable at 121ºC (250ºF).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty