Would you like to log in again?
PYREX® Burets, Class B, Straight Bore Fluoropolymer Resin Stopcock, Colored Scale, Corning
Supplier: Corning
PYREX®
2122A-10 2122A-100 2122A-25 2122A-50
17452-164CS 279.29 USD InStock
17452-164 17452-120 17452-142 17452-109
PYREX® Burets, Class B, Straight Bore Fluoropolymer Resin Stopcock, Colored Scale, Corning
Burets
Borosilicate glass.
  • Microfinish of the barrel provides a precision fit
  • Durable colored markings
These burets are suitable for use in school and institutional laboratories where the accuracy of class A models is not required.
The tips are carefully drawn for accuracy to insure proper drainage rate.
Delivery information: Burets are supplied with a 2 mm PTFE stopcock plug and dust cover.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty