Would you like to log in again?
PYREX® Burets, Serialized and Certified, Class A, Precision Bore, Straight [ST] Stopcock, Corning
Supplier: Corning
PYREX®
2135-100 2135-50 2135-10 2135-25
17473-087CS 417.37 USD InStock
17473-087 17473-096 17473-090 17473-093
PYREX® Burets, Serialized and Certified, Class A, Precision Bore, Straight [ST] Stopcock, Corning
Burets
PYREX® borosilicate glass, class A.
  • Durable white graduations, sharply defined
  • Easy to read
These precision bore tubing is used to assure accuracy between any two points. Each buret is individually serialized and supplied with a certificate of identification and capacity, traceable to NIST standards.
Capacity tolerance is established by ASTM E-542 and ASTM E-287
PYREX® Serialized/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Durable colored graduations are sharply defined and easy-to-read. Carefully ground [ST] stopcock gives precise control.
Delivery information: Burets are supplied with bore stopcock plug and dust cover.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty